Assay Details
Target Gene Details
Entrez Gene ID: | 57212 |
Gene Name: | TP73 antisense RNA 1 |
Gene Aliases: |
KIAA0495, PDAM |
Location: |
Chr.1:3735984-3747373 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TP73-AS1 | NR_033708.1 | 5 | 3449 | |
NR_033709.1 | 5 | 4062 | ||
NR_033710.1 | 5 | 5317 | ||
NR_033711.1 | 4 | 5781 | ||
NR_033712.1 | 6 | 2696 | ||
AB007964.1 | 4 | 5731 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470687 | Chr.1:3356499 - 3763210 on Build GRCh38 | Loss | MIR551A TP73-AS1 WRAP73 TP73 TPRG1L PRDM16 ARHGEF16 CCDC27 MEGF6 |
nsv460061 | Chr.1:3680453 - 3755227 on Build GRCh38 | Loss | TP73-AS1 TP73 CCDC27 |
esv994567 | Chr.1:3735559 - 3738806 on Build GRCh38 | Deletion | TP73-AS1 TP73 |
nsv1001715 | Chr.1:3627619 - 3769754 on Build GRCh38 | Loss | TP73-AS1 WRAP73 TP73 TPRG1L CCDC27 |
esv3585042 | Chr.1:3585575 - 4260083 on Build GRCh38 | Gain | TP73-AS1 LINC01134 WRAP73 TP73 TPRG1L LRRC47 CCDC27 SMIM1 LINC01346 CEP104 C1orf174 DFFB MEGF6 |
nsv1012423 | Chr.1:3477686 - 4747449 on Build GRCh38 | Gain | TP73-AS1 LINC01134 WRAP73 TP73 TPRG1L LRRC47 CCDC27 LOC284661 AJAP1 SMIM1 LINC01346 MIR551A CEP104 C1orf174 DFFB ARHGEF16 MEGF6 |
nsv951959 | Chr.1:3051037 - 3812036 on Build GRCh38 | Deletion | TP73-AS1 WRAP73 TP73 TPRG1L PRDM16 LRRC47 MIR4251 CCDC27 SMIM1 MIR551A LINC00982 LOC105378604 ARHGEF16 MEGF6 |
nsv830314 | Chr.1:3660776 - 3804275 on Build GRCh38 | Gain+Loss | TP73-AS1 TP73 LRRC47 CCDC27 SMIM1 |
dgv13n100 | Chr.1:3730448 - 3791316 on Build GRCh38 | Gain | TP73-AS1 TP73 LRRC47 CCDC27 SMIM1 |
esv2758916 | Chr.1:3668477 - 3738972 on Build GRCh38 | Gain | TP73-AS1 TP73 |