Assay Details
Target Gene Details
Entrez Gene ID: | 730102 |
Gene Name: | quinone oxidoreductase-like protein 2 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.1:178006136-178038007 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC730102 | NR_037167.1 | |||
BX538051.1 | ||||
DA045540.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv548252 | Chr.1:177758922 - 178428784 on Build GRCh38 | Gain | LOC730102 SEC16B RASAL2 RASAL2-AS1 |
nsv523131 | Chr.1:178011924 - 178031386 on Build GRCh38 | Loss | LOC730102 |