Assay Details
Target Gene Details
Entrez Gene ID: | 55870 |
Gene Name: | ASH1 like histone lysine methyltransferase |
Gene Aliases: |
ASH1, ASH1L1, KMT2H |
Location: |
Chr.1:155335261-155563160 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASH1L | NM_018489.2 | NP_060959.2 | ||
XM_006711450.3 | XP_006711513.1 | |||
XM_006711451.3 | XP_006711514.1 | |||
XM_006711452.3 | XP_006711515.1 | |||
XM_011509770.2 | XP_011508072.1 | |||
XM_017001784.1 | XP_016857273.1 | |||
XM_017001785.1 | XP_016857274.1 | |||
XM_017001788.1 | XP_016857277.1 | |||
AF257305.1 | AAF68983.1 | |||
AK304873.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831636 | Chr.1:155249432 - 155475073 on Build GRCh38 | Loss | LOC105371451 HCN3 FAM189B FDPS ASH1L RUSC1-AS1 RUSC1 POU5F1P4 SCAMP3 PKLR CLK2 MIR555 |
esv33869 | Chr.1:155253492 - 155948170 on Build GRCh38 | Loss | HCN3 GON4L LOC100505728 SNORA80E RUSC1 POU5F1P4 SCAMP3 SYT11 FAM189B SCARNA26B ASH1L RUSC1-AS1 SCARNA4 PKLR RIT1 CLK2 RXFP4 KIAA0907 MSTO2P YY1AP1 ARHGEF2 SCARNA26A LOC105371451 DAP3 FDPS ASH1L-AS1 MSTO1 MIR555 |
dgv3n68 | Chr.1:155122502 - 155343618 on Build GRCh38 | Loss | HCN3 MUC1 GBAP1 MTX1 RUSC1 SLC50A1 SCAMP3 MIR92B LOC105371451 EFNA1 FAM189B FDPS ASH1L RUSC1-AS1 GBA TRIM46 PKLR THBS3 CLK2 KRTCAP2 DPM3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |