Assay Details
Target Gene Details
Entrez Gene ID: | 100288960 |
Gene Name: | protease, serine 43 |
Gene Aliases: |
TESSP3 |
Location: |
Chr.3:46753043-46812690 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRSS43 | XM_017007617.1 | XP_016863106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2641432 | Chr.3:46755646 - 46807786 on Build GRCh38 | Loss |
![]() |
esv26141 | Chr.3:46752934 - 46810030 on Build GRCh38 | Loss |
![]() |
esv3893692 | Chr.3:46754860 - 46809120 on Build GRCh38 | Loss |
![]() |
nsv498998 | Chr.3:46754014 - 46809829 on Build GRCh38 | Loss |
![]() |
nsv965407 | Chr.3:46754541 - 46808658 on Build GRCh38 | Deletion |
![]() |
esv2725238 | Chr.3:46753526 - 46809261 on Build GRCh38 | Deletion |
![]() |
nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv514149 | Chr.3:46753234 - 46808734 on Build GRCh38 | Loss |
![]() |
dgv102e203 | Chr.3:46747685 - 46838512 on Build GRCh38 | Loss |
![]() ![]() |
dgv31n50 | Chr.3:46748907 - 46811189 on Build GRCh38 | Loss |
![]() |
nsv438397 | Chr.3:46756350 - 46805202 on Build GRCh38 | Loss |
![]() |
esv2421845 | Chr.3:46755299 - 46808099 on Build GRCh38 | Deletion |
![]() |
nsv10269 | Chr.3:46752614 - 46810227 on Build GRCh38 | Loss |
![]() |
dgv1372e212 | Chr.3:46751582 - 46818593 on Build GRCh38 | Loss |
![]() |
esv2488989 | Chr.3:46753480 - 46810454 on Build GRCh38 | Deletion |
![]() |
nsv516623 | Chr.3:46638573 - 46811189 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
dgv4734n100 | Chr.3:46751875 - 46818330 on Build GRCh38 | Gain+Loss |
![]() |
nsv1112826 | Chr.3:46754110 - 46771910 on Build GRCh38 | Deletion |
![]() |
nsv1160935 | Chr.3:46751887 - 46808099 on Build GRCh38 | Deletion |
![]() |
dgv4731n100 | Chr.3:46733893 - 46827326 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv4730n100 | Chr.3:46729944 - 46808087 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv8305n54 | Chr.3:46755146 - 46779343 on Build GRCh38 | Loss |
![]() |
esv32999 | Chr.3:46754570 - 46808583 on Build GRCh38 | Gain+Loss |
![]() |
dgv4732n100 | Chr.3:46750043 - 46838650 on Build GRCh38 | Loss |
![]() ![]() |
dgv860e199 | Chr.3:46753848 - 46809682 on Build GRCh38 | Deletion |
![]() |
nsv3798 | Chr.3:46737140 - 46824116 on Build GRCh38 | Deletion |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)