Assay Details
Target Gene Details
Entrez Gene ID: | 26472 |
Gene Name: | protein phosphatase 1 regulatory inhibitor subunit 14B |
Gene Aliases: |
PHI-1, PLCB3N, PNG, SOM172 |
Location: |
Chr.11:64244479-64246941 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP1R14B | NM_138689.2 | NP_619634.1 | ||
AF318364.1 | AAL55871.1 | |||
BC014522.2 | AAH14522.3 | |||
BC094817.1 | AAH94817.1 | |||
BF155230.1 | ||||
BT009914.1 | AAP88916.1 | |||
X91195.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1159904 | Chr.11:64150129 - 64279169 on Build GRCh38 | Deletion | BAD LOC105369340 DNAJC4 PPP1R14B STIP1 VEGFB TRPT1 PLCB3 GPR137 MACROD1 FKBP2 NUDT22 FERMT3 |
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | TEX40 TRMT112 BAD LOC105369340 KCNK4 PPP1R14B STIP1 COX8A PLCB3 MACROD1 OTUB1 FLRT1 FKBP2 FERMT3 KCNK4-TEX40 PRDX5 DNAJC4 VEGFB MIR7155 TRPT1 GPR137 NAA40 CCDC88B ESRRA NUDT22 |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss | TEX40 KCNK4-TEX40 TRMT112 BAD LOC105369340 PRDX5 DNAJC4 KCNK4 PPP1R14B STIP1 VEGFB TRPT1 PLCB3 GPR137 FKBP2 ESRRA NUDT22 FERMT3 |
nsv555187 | Chr.11:64185649 - 64260416 on Build GRCh38 | Loss | DNAJC4 PPP1R14B STIP1 VEGFB TRPT1 PLCB3 FKBP2 LOC105369340 NUDT22 FERMT3 |
nsv983032 | Chr.11:64243929 - 64247666 on Build GRCh38 | Duplication | PPP1R14B FKBP2 LOC105369340 |
esv24861 | Chr.11:64233793 - 64250642 on Build GRCh38 | Gain | DNAJC4 PPP1R14B VEGFB FKBP2 LOC105369340 |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss | TEX40 KCNK4-TEX40 TRMT112 BAD LOC105369340 MIR1237 PRDX5 DNAJC4 KCNK4 PPP1R14B VEGFB MIR7155 RPS6KA4 TRPT1 PLCB3 GPR137 CCDC88B FKBP2 ESRRA NUDT22 FERMT3 |
nsv555184 | Chr.11:64156974 - 64275525 on Build GRCh38 | Loss | BAD LOC105369340 DNAJC4 PPP1R14B STIP1 VEGFB TRPT1 PLCB3 GPR137 MACROD1 FKBP2 NUDT22 FERMT3 |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss | TEX40 KCNK4-TEX40 TRMT112 BAD LOC105369340 PRDX5 DNAJC4 KCNK4 PPP1R14B STIP1 VEGFB TRPT1 PLCB3 GPR137 MACROD1 FLRT1 FKBP2 ESRRA NUDT22 FERMT3 |
More Information
Additional Information:
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation Validated |