Assay Details
Target Gene Details
Entrez Gene ID: | 730100 |
Gene Name: | uncharacterized LOC730100 |
Gene Aliases: |
- |
Location: |
Chr.2:51032601-52407917 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC730100 | NR_135237.1 | |||
AK127244.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1000086 | Chr.2:50879876 - 51193400 on Build GRCh38 | Loss | NRXN1 LOC730100 |
esv2759048 | Chr.2:51054576 - 51138744 on Build GRCh38 | Gain | LOC730100 |
nsv2725 | Chr.2:51046194 - 51067339 on Build GRCh38 | Insertion | LOC730100 |
nsv581868 | Chr.2:51053379 - 51125305 on Build GRCh38 | Loss | LOC730100 |
nsv521850 | Chr.2:51052256 - 51210278 on Build GRCh38 | Loss | LOC730100 |
dgv3814n100 | Chr.2:51044124 - 51123977 on Build GRCh38 | Loss | LOC730100 |
nsv1012208 | Chr.2:51039660 - 51188856 on Build GRCh38 | Loss | LOC730100 |
esv3892237 | Chr.2:51020156 - 51130764 on Build GRCh38 | Loss | NRXN1 LOC730100 |
dgv6866n54 | Chr.2:50820175 - 51108720 on Build GRCh38 | Loss | NRXN1 LOC730100 |
nsv581848 | Chr.2:50711821 - 51118377 on Build GRCh38 | Loss | NRXN1 LOC730100 |
nsv581853 | Chr.2:50786509 - 51326909 on Build GRCh38 | Loss | NRXN1 LOC730100 |
esv2763321 | Chr.2:50610356 - 51126248 on Build GRCh38 | Loss | NRXN1 MIR8485 LOC730100 |
esv3892215 | Chr.2:50887566 - 51133529 on Build GRCh38 | Loss | NRXN1 LOC730100 |
nsv834014 | Chr.2:50954708 - 51133399 on Build GRCh38 | Loss | NRXN1 LOC730100 |
nsv1002086 | Chr.2:50685023 - 51177923 on Build GRCh38 | Loss | NRXN1 MIR8485 LOC730100 |