Assay Details
Target Gene Details
Entrez Gene ID: | 161931 |
Gene Name: | adenosine deaminase domain containing 2 |
Gene Aliases: |
TENRL |
Location: |
Chr.16:84191117-84197166 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADAD2 | NM_001145400.1 | NP_001138872.1 | ||
NM_139174.3 | NP_631913.3 | |||
XM_005255814.1 | XP_005255871.1 | |||
XM_011522919.1 | XP_011521221.1 | |||
XM_011522920.1 | XP_011521222.1 | |||
AF447586.1 | AAM22869.1 | |||
AK093046.1 | BAC04032.1 | |||
AK093523.1 | ||||
AK131102.1 | BAC85152.1 | |||
AK315171.1 | ||||
BC033491.1 | AAH33491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3639413 | Chr.16:84172318 - 84253359 on Build GRCh38 | Loss | ADAD2 DNAAF1 KCNG4 TAF1C |
nsv518342 | Chr.16:84194593 - 84196629 on Build GRCh38 | Loss | ADAD2 |
nsv833309 | Chr.16:84076867 - 84248591 on Build GRCh38 | Loss | LOC729887 MBTPS1 ADAD2 DNAAF1 KCNG4 HSDL1 TAF1C |
nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss | LOC729887 MBTPS1 OSGIN1 ADAD2 MLYCD LOC105371371 DNAAF1 KCNG4 HSDL1 TAF1C SLC38A8 NECAB2 |
esv2761930 | Chr.16:84007420 - 84312461 on Build GRCh38 | Gain | LOC729887 MBTPS1 LOC105376777 ADAD2 DNAAF1 KCNG4 HSDL1 WFDC1 TAF1C SLC38A8 |
nsv523437 | Chr.16:84176132 - 84196770 on Build GRCh38 | Loss | ADAD2 DNAAF1 TAF1C |
esv3639414 | Chr.16:84176251 - 84352273 on Build GRCh38 | Loss | LOC105376777 ADAD2 DNAAF1 KCNG4 WFDC1 TAF1C |
nsv457597 | Chr.16:84182086 - 84215151 on Build GRCh38 | Loss | ADAD2 TAF1C |
esv3639412 | Chr.16:84170074 - 84221406 on Build GRCh38 | Loss | ADAD2 DNAAF1 KCNG4 TAF1C |
nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain | FOXC2 LINC00917 USP10 LOC101928557 DNAAF1 KLHL36 LINC01082 FOXL1 LOC105376777 ADAD2 MIR5093 EMC8 COTL1 MTHFSD FAM92B KCNG4 CRISPLD2 LOC400548 FLJ30679 WFDC1 IRF8 FOXF1 MIR1910 LOC101928474 COX4I1 LINC00311 LOC102724344 FENDRR TAF1C GINS2 ZDHHC7 MIR6774 KIAA0513 LOC146513 FOXC2-AS1 TLDC1 LOC101928614 LOC440390 LINC01081 LOC105371374 GSE1 ATP2C2 C16orf74 |
nsv1059227 | Chr.16:84029323 - 84233403 on Build GRCh38 | Loss | LOC729887 MBTPS1 ADAD2 DNAAF1 KCNG4 HSDL1 TAF1C SLC38A8 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |