Assay Details
Target Gene Details
Entrez Gene ID: | 253152 |
Gene Name: | epoxide hydrolase 4 |
Gene Aliases: |
ABHD7, EH4, EPHXRP |
Location: |
Chr.1:92029976-92063536 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHX4 | NM_173567.4 | 6 | 903 | NP_775838.3 |
AK074822.1 | 6 | 863 | BAC11230.1 | |
BC041475.1 | 6 | 878 | AAH41475.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1002730 | Chr.1:91836984 - 92099672 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv524845 | Chr.1:91750168 - 92133148 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv2750842 | Chr.1:91891224 - 92114118 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1855 | Chr.1:92045254 - 92075776 on Build GRCh38 | Insertion |
![]() ![]() |
nsv1006318 | Chr.1:91750851 - 92142767 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)