Assay Details
Target Gene Details
Entrez Gene ID: | 1740 |
Gene Name: | discs large MAGUK scaffold protein 2 |
Gene Aliases: |
PPP1R58, PSD-93, PSD93, chapsyn-110 |
Location: |
Chr.11:83455009-85628534 on Build GRCh38 |
Assay Gene Location: | Within Exon 43 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33531 | Chr.11:82520885 - 83805659 on Build GRCh38 | Loss | SNORA70E DDIAS PRCP LOC102724712 PCF11-AS1 ANKRD42 PCF11 RAB30-AS1 DLG2 FAM181B LOC100506282 RAB30 CCDC90B |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |