Assay Details
Target Gene Details
Entrez Gene ID: | 4696 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A3 |
Gene Aliases: |
B9, CI-B9 |
Location: |
Chr.19:54102338-54106974 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFA3 | NM_004542.3 | NP_004533.1 | ||
XM_017026833.1 | XP_016882322.1 | |||
AF044955.1 | AAD05420.1 | |||
AF070653.1 | AAD20959.1 | |||
AK289489.1 | ||||
BC022369.1 | AAH22369.1 | |||
BC061644.1 | AAH61644.1 | |||
CR542143.1 | CAG46940.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1056177 | Chr.19:54073048 - 54112306 on Build GRCh38 | Gain | TARM1 TFPT OSCAR NDUFA3 |
nsv510773 | Chr.19:54017180 - 54132236 on Build GRCh38 | Deletion | TARM1 TFPT OSCAR VSTM1 NDUFA3 PRPF31 |
nsv515623 | Chr.19:54102901 - 54179150 on Build GRCh38 | Loss | MBOAT7 LENG1 CNOT3 TFPT TMC4 NDUFA3 PRPF31 |
nsv953613 | Chr.19:54097214 - 54198733 on Build GRCh38 | Deletion | MBOAT7 LENG1 TSEN34 CNOT3 TFPT OSCAR TMC4 NDUFA3 PRPF31 |
esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion | LENG1 MIR517B MIR1283-2 MIR372 TARM1 TFPT BIRC8 VN1R2 LOC105372457 LILRA5 MIR523 MIR371A VN1R4 FAM90A27P MYADM MIR520B MIR519B MIR498 MIR520G MIR512-1 CACNG8 ZNF813 ZNF665 LILRB2 NDUFA3 MIR524 MIR1283-1 MIR526A2 MIR516B2 MIR519D MIR515-1 RPS9 PRKCG LOC284379 MIR520A ZNF818P MIR519C MIR512-2 MIR520C NLRP12 PRPF31 ZNF677 MIR521-1 MIR518D TSEN34 MIR526A1 ZNF845 MIR516B1 MIR519E MIR518B ZNF761 VSTM1 MIR527 ZNF765 MIR935 MIR519A2 TMC4 MIR521-2 CACNG6 MIR520D LILRA6 CACNG7 MIR522 MIR520H MIR518A2 MIR518C MIR520E MIR1323 OSCAR MIR515-2 MIR518F LILRB5 MIR516A1 MIR526B MIR520F ZNF525 MBOAT7 MIR516A2 CNOT3 MIR373 MIR518E MIR4752 MIR518A1 MIR519A1 TPM3P9 MIR517C MIR517A ZNF331 MIR525 MIR371B LILRB3 DPRX |
nsv470157 | Chr.19:54063999 - 54127773 on Build GRCh38 | Loss | TARM1 TFPT OSCAR NDUFA3 PRPF31 |
esv3893246 | Chr.19:54081109 - 54111207 on Build GRCh38 | Gain | TARM1 TFPT OSCAR NDUFA3 |
nsv833877 | Chr.19:53951529 - 54117323 on Build GRCh38 | Loss | TARM1 CACNG8 TFPT OSCAR VSTM1 MIR935 NDUFA3 CACNG6 PRPF31 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |