Assay Details
Target Gene Details
Entrez Gene ID: | 56889 |
Gene Name: | transmembrane 9 superfamily member 3 |
Gene Aliases: |
EP70-P-iso, SMBP |
Location: |
Chr.10:96518110-96587052 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TM9SF3 | NM_020123.3 | 15 | 2114 | NP_064508.3 |
XM_011539976.2 | 15 | 2194 | XP_011538278.1 | |
XM_011539977.1 | 15 | 2243 | XP_011538279.1 | |
AF160213.1 | 15 | 1907 | AAF67014.1 | |
AF269150.1 | 15 | 2114 | AAF98159.1 | |
AK000756.1 | 13 | 1517 | BAA91362.1 | |
AK027438.1 | 10 | 1164 | BAB55110.1 | |
AK027788.1 | 14 | 1770 | BAB55369.1 | |
AK074824.1 | 12 | 1405 | BAC11232.1 | |
AK075093.1 | 10 | 1152 | BAC11397.1 | |
AK075451.1 | 4 | 517 | ||
AK297721.1 | ||||
AY358819.1 | 14 | 1784 | AAQ89178.1 | |
BC020959.1 | 14 | 1694 | AAH20959.2 | |
BC136788.1 | 15 | 2114 | ||
BC136789.1 | 15 | 2114 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3577627 | Chr.10:96515251 - 96710544 on Build GRCh38 | Gain | TM9SF3 PIK3AP1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78610910] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic DGV Variation |