Assay Details
Target Gene Details
Entrez Gene ID: | 4850 |
Gene Name: | CCR4-NOT transcription complex subunit 4 |
Gene Aliases: |
CLONE243, NOT4, NOT4H |
Location: |
Chr.7:135361795-135510127 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CNOT4 | NM_001190847.1 | 11 | 2430 | NP_001177776.1 |
NM_001190849.1 | 12 | 2634 | NP_001177778.1 | |
NM_001190850.1 | 12 | 2643 | NP_001177779.1 | |
NM_013316.3 | 11 | 2421 | NP_037448.2 | |
XM_017012235.1 | 12 | 2504 | XP_016867724.1 | |
AF091094.1 | 8 | 1695 | AAC72963.1 | |
AK074493.1 | ||||
AK074671.1 | 10 | 1830 | BAC11125.1 | |
AK300365.1 | 12 | 2600 | ||
U71267.1 | 11 | 2380 | AAD00179.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv29990 | Chr.7:135328379 - 135372394 on Build GRCh38 | Gain | CNOT4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |