Assay Details
Target Gene Details
Entrez Gene ID: | 5610 |
Gene Name: | eukaryotic translation initiation factor 2 alpha kinase 2 |
Gene Aliases: |
EIF2AK1, PKR, PPP1R83, PRKR |
Location: |
Chr.2:37099210-37157065 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF2AK2 | NM_001135651.2 | 15 | 1749 | NP_001129123.1 |
NM_001135652.2 | 12 | 1319 | NP_001129124.1 | |
NM_002759.3 | 15 | 1983 | NP_002750.1 | |
XM_011532987.2 | 15 | 1683 | XP_011531289.1 | |
XM_017004503.1 | 14 | 1550 | XP_016859992.1 | |
AF086566.1 | 3 | 354 | ||
AK290655.1 | 15 | 1669 | ||
AK313818.1 | 14 | 1506 | ||
AY302136.1 | 12 | 1303 | AAP57628.1 | |
BC040851.1 | 15 | 1749 | AAH40851.1 | |
BC057805.1 | 15 | 1654 | AAH57805.1 | |
BC093676.1 | 13 | 1454 | AAH93676.1 | |
BC101475.1 | 13 | 1454 | AAI01476.1 | |
BC143230.1 | 13 | 1454 | ||
BM473760.1 | 2 | 96 | ||
M35663.1 | 14 | 1612 | AAA36409.1 | |
M85294.1 | 13 | 1456 | AAA18253.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |