Assay Details
Target Gene Details
Entrez Gene ID: | 7163 |
Gene Name: | tumor protein D52 |
Gene Aliases: |
D52, N8L, PC-1, PrLZ, hD52 |
Location: |
Chr.8:80034868-80171659 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TPD52 | NM_001025252.2 | 6 | 1527 | NP_001020423.1 |
NM_001025253.2 | 8 | 1314 | NP_001020424.1 | |
NM_001287140.1 | 8 | 1596 | NP_001274069.1 | |
NM_001287142.1 | 7 | 1569 | NP_001274071.1 | |
NM_001287143.1 | 7 | 1554 | NP_001274072.1 | |
NM_001287144.1 | 6 | 1238 | NP_001274073.1 | |
NM_005079.3 | 6 | 1245 | NP_005070.1 | |
NR_105033.1 | 9 | 1666 | ||
NR_105034.1 | 5 | 1129 | ||
NR_105035.1 | 7 | 1315 | ||
NR_105036.1 | 8 | 1259 | ||
NR_105037.1 | 7 | 1260 | ||
AF202897.1 | 6 | 1471 | AAO22156.1 | |
AK057075.1 | 8 | 1315 | ||
AK223631.1 | BAD97351.1 | |||
AK295468.1 | 8 | 1181 | ||
AK296615.1 | 6 | 1249 | ||
AK308983.1 | 8 | 1171 | ||
BC018117.1 | 6 | 1116 | AAH18117.1 | |
BX640835.1 | 6 | 1311 | CAE45908.1 | |
GQ499324.1 | 8 | 1581 | ||
GQ499325.1 | 7 | 1554 | ||
GQ499326.1 | 7 | 1539 | ||
GQ499327.1 | 6 | 1512 | ||
GQ499328.1 | 9 | 1651 | ||
S82081.1 | 6 | 1122 | AAB36475.1 | |
U18914.1 | 6 | 1120 | AAC50183.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |