Assay Details
Target Gene Details
Entrez Gene ID: | 2017 |
Gene Name: | cortactin |
Gene Aliases: |
EMS1 |
Location: |
Chr.11:70398506-70436584 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CTTN | NM_001184740.1 | 5 | 439 | NP_001171669.1 |
NM_005231.3 | 5 | 439 | NP_005222.2 | |
NM_138565.2 | 5 | 439 | NP_612632.1 | |
XM_006718447.2 | 5 | 439 | XP_006718510.1 | |
XM_006718448.3 | 5 | 439 | XP_006718511.1 | |
XM_017017312.1 | 5 | 439 | XP_016872801.1 | |
AK222613.1 | 5 | 408 | BAD96333.1 | |
AK291097.1 | 5 | 410 | ||
AU124856.1 | 5 | 407 | ||
BC008799.2 | 5 | 401 | AAH08799.1 | |
BC033889.1 | 5 | 409 | AAH33889.1 | |
DA716772.1 | 5 | 439 | ||
M98343.1 | 5 | 401 | AAA58455.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |