Assay Details
Target Gene Details
Entrez Gene ID: | 286183 |
Gene Name: | Na+/K+ transporting ATPase interacting 3 |
Gene Aliases: |
FAM77D |
Location: |
Chr.8:62248942-62999652 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NKAIN3 | NR_130764.1 | |||
XM_017013359.1 | XP_016868848.1 | |||
AK096949.1 | BAC04909.1 |
Target Gene Details
Entrez Gene ID: | 643763 |
Gene Name: | uncharacterized LOC643763 |
Gene Aliases: |
- |
Location: |
Chr.8:62977861-62984901 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UG0898H09 | NR_027378.1 | 1 | 2914 | |
AF360117.1 | 1 | 2914 | AAN76520.1 | |
AK096517.1 | 1 | 2268 | BAC04808.1 | |
AK127279.1 | 1 | 2829 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1018879 | Chr.8:62369165 - 63025073 on Build GRCh38 | Gain | UG0898H09 NKAIN3 GGH |
nsv6227 | Chr.8:62979528 - 63024393 on Build GRCh38 | Deletion | UG0898H09 NKAIN3 GGH |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |