Assay Details
Target Gene Details
Entrez Gene ID: | 9557 |
Gene Name: | chromodomain helicase DNA binding protein 1 like |
Gene Aliases: |
ALC1, CHDL |
Location: |
Chr.1:147172804-147295766 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHD1L | XM_017002858.1 | XP_016858347.1 |
Target Gene Details
Entrez Gene ID: | 2330 |
Gene Name: | flavin containing monooxygenase 5 |
Gene Aliases: |
- |
Location: |
Chr.1:147183963-147225798 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FMO5 | NM_001144829.2 | 6 | 1195 | NP_001138301.1 |
NM_001144830.2 | 5 | 1405 | NP_001138302.1 | |
NM_001461.3 | 6 | 1185 | NP_001452.2 | |
XM_005272946.4 | 6 | 1159 | XP_005273003.1 | |
XM_005272947.4 | 6 | 903 | XP_005273004.1 | |
XM_005272948.4 | 6 | 886 | XP_005273005.1 | |
XM_006711244.3 | 6 | 1175 | XP_006711307.1 | |
XM_006711245.3 | 6 | 1175 | XP_006711308.1 | |
XM_011509350.2 | 6 | 1190 | XP_011507652.1 | |
XM_011509351.2 | 5 | 922 | XP_011507653.1 | |
XM_017000801.1 | 5 | 934 | XP_016856290.1 | |
XM_017000802.1 | 5 | 704 | XP_016856291.1 | |
AK022172.1 | 5 | 1395 | BAB13975.1 | |
AK222728.1 | 6 | 900 | BAD96448.1 | |
AK225739.1 | 6 | 910 | ||
AK314647.1 | 6 | 1113 | ||
BC035687.1 | 5 | 1078 | AAH35687.1 | |
L37080.1 | 6 | 871 | AAA67849.1 | |
Z47553.1 | 6 | 877 | CAA87633.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv327n100 | Chr.1:146028214 - 148860637 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1005055 | Chr.1:145642988 - 148991068 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2761299 | Chr.1:146068341 - 148926801 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv509457 | Chr.1:145948024 - 149021821 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1132304 | Chr.1:146186094 - 148977674 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1003688 | Chr.1:146036817 - 148934526 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv66e199 | Chr.1:146055970 - 149056514 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv437047 | Chr.1:147171976 - 147369886 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)