Assay Details
Target Gene Details
Entrez Gene ID: | 10138 |
Gene Name: | YY1 associated factor 2 |
Gene Aliases: |
- |
Location: |
Chr.12:42157104-42238368 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YAF2 | NM_001190977.2 | 3 | 1992 | NP_001177906.1 |
NM_001190979.2 | 5 | 2190 | NP_001177908.1 | |
NM_001320080.1 | 5 | 2218 | NP_001307009.1 | |
NM_005748.5 | 4 | 2118 | NP_005739.2 | |
NR_135139.1 | 7 | 2360 | ||
NR_135140.1 | 7 | 2392 | ||
XM_006719184.3 | 4 | 2037 | XP_006719247.1 | |
XM_006719185.3 | 4 | 2062 | XP_006719248.1 | |
XM_011537728.2 | 6 | 2217 | XP_011536030.1 | |
XM_011537729.2 | 4 | 2007 | XP_011536031.1 | |
XM_017018669.1 | 6 | 2176 | XP_016874158.1 | |
BC037777.2 | 5 | 2058 | AAH37777.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |