Assay Details
Target Gene Details
Entrez Gene ID: | 56949 |
Gene Name: | XPA binding protein 2 |
Gene Aliases: |
HCNP, HCRN, NTC90, SYF1 |
Location: |
Chr.19:7619525-7629553 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XAB2 | NM_020196.2 | NP_064581.2 | ||
AB026111.1 | BAB15807.1 | |||
AB033003.1 | BAA86491.1 | |||
AF226051.1 | AAF86951.1 | |||
AF258567.1 | AAG23770.1 | |||
AF272147.1 | AAQ14304.1 | |||
AK025858.1 | ||||
AK074035.1 | BAB84861.1 | |||
BC007208.1 | AAH07208.1 | |||
BC008778.1 | AAH08778.1 | |||
CR749864.1 | CAH18708.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953958 | Chr.19:7584715 - 7650814 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv833731 | Chr.19:7589114 - 7739331 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1060887 | Chr.19:7089546 - 7861087 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv993190 | Chr.19:7615627 - 7629366 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv833730 | Chr.19:7509505 - 7633597 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2751809 | Chr.19:6896369 - 7816144 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)