Assay Details
Target Gene Details
Entrez Gene ID: | 10930 |
Gene Name: | apolipoprotein B mRNA editing enzyme catalytic subunit 2 |
Gene Aliases: |
ARCD1, ARP1 |
Location: |
Chr.6:41053201-41064891 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOBEC2 | NM_006789.3 | 2 | 623 | NP_006780.1 |
AF161698.1 | 2 | 531 | AAD45360.1 | |
AK223461.1 | 2 | 623 | BAD97181.1 | |
AK300689.1 | 3 | 397 | ||
AK308197.1 | 2 | 454 | ||
AK310006.1 | ||||
AK313288.1 | 2 | 624 | ||
BC047767.2 | 2 | 560 | AAH47767.1 | |
BC069688.1 | 2 | 520 | AAH69688.1 | |
BC069764.1 | 2 | 523 | AAH69764.1 | |
BI438931.1 | ||||
CB051273.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |