Assay Details
Target Gene Details
Entrez Gene ID: | 91624 |
Gene Name: | nexilin F-actin binding protein |
Gene Aliases: |
CMH20, NELIN |
Location: |
Chr.1:77888515-77946329 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NEXN | NM_001172309.1 | 4 | 487 | NP_001165780.1 |
NM_144573.3 | 5 | 679 | NP_653174.3 | |
XM_005271322.3 | 5 | 570 | XP_005271379.1 | |
XM_005271323.3 | 5 | 570 | XP_005271380.1 | |
XM_005271324.4 | 4 | 474 | XP_005271381.1 | |
XM_005271325.3 | 5 | 570 | XP_005271382.1 | |
XM_005271326.3 | 4 | 405 | XP_005271383.1 | |
XM_005271327.3 | 5 | 570 | XP_005271384.1 | |
AF114264.1 | 1 | 109 | AAD29607.1 | |
AF176780.1 | 4 | 358 | AAQ13658.1 | |
AK057954.1 | 1 | 87 | BAB71622.1 | |
AK298565.1 | 4 | 456 | ||
AK300728.1 | 5 | 612 | ||
AK307487.1 | 4 | 405 | ||
AY211912.1 | 5 | 532 | AAO65165.1 | |
BC017827.1 | ||||
BC055084.1 | 5 | 456 | AAH55084.1 | |
BC111395.1 | 2 | 110 | ||
BC114444.1 | 1 | 85 | ||
BC114445.1 | 1 | 85 | ||
BX648546.1 | 5 | 567 | ||
CR936630.1 | 5 | 589 | CAI56773.1 | |
DC412178.1 | 4 | 488 | ||
DQ464902.1 | 5 | 566 | ||
S67069.1 | 1 | 61 | AAB28815.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |