Assay Details
Target Gene Details
Entrez Gene ID: | 5156 |
Gene Name: | platelet derived growth factor receptor alpha |
Gene Aliases: |
CD140A, GAS9, PDGFR-2, PDGFR2, RHEPDGFRA |
Location: |
Chr.4:54229097-54298245 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDGFRA | NM_006206.4 | 7 | 1347 | NP_006197.1 |
XM_005265743.1 | 7 | 1131 | XP_005265800.1 | |
XM_006714039.3 | 8 | 1392 | XP_006714102.1 | |
XM_006714041.3 | 8 | 1395 | XP_006714104.1 | |
XM_011534385.2 | 7 | 1385 | XP_011532687.1 | |
XM_017008280.1 | 7 | 1562 | XP_016863769.1 | |
XM_017008281.1 | 7 | 1628 | XP_016863770.1 | |
XM_017008282.1 | 7 | 1276 | XP_016863771.1 | |
AK308353.1 | 7 | 1177 | ||
AK311006.1 | 6 | 834 | ||
AK316578.1 | 7 | 1149 | ||
BC063414.1 | 7 | 1131 | AAH63414.1 | |
M21574.1 | 7 | 1155 | AAA96715.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77524207] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |