Assay Details
Target Gene Details
Entrez Gene ID: | 50640 |
Gene Name: | patatin like phospholipase domain containing 8 |
Gene Aliases: |
IPLA2-2, IPLA2G, MMLA, PNPLA-gamma, iPLA2gamma |
Location: |
Chr.7:108470417-108528161 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PNPLA8 | NM_001256007.2 | 4 | 1523 | NP_001242936.1 |
NM_001256008.2 | 3 | 1477 | NP_001242937.1 | |
NM_001256009.2 | 3 | 1477 | NP_001242938.1 | |
NM_001256010.2 | 3 | 1350 | NP_001242939.1 | |
NM_001256011.2 | 4 | 1211 | NP_001242940.1 | |
NM_015723.4 | 5 | 1635 | NP_056538.1 | |
XM_005250396.4 | 4 | 2479 | XP_005250453.1 | |
XM_011516274.2 | 4 | 2352 | XP_011514576.1 | |
XM_011516275.2 | 4 | 2352 | XP_011514577.1 | |
XM_017012261.1 | 3 | 2306 | XP_016867750.1 | |
AB041261.1 | 3 | 1303 | BAA94997.1 | |
AF263613.1 | 4 | 1329 | AAF75269.1 | |
AK024335.1 | 4 | 1211 | BAB14890.1 | |
AK291755.1 | 4 | 1340 | ||
AL834147.1 | 3 | 1230 | CAD38859.1 | |
BC032999.2 | 5 | 1511 | AAH32999.1 | |
BX647865.1 | 3 | 1196 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |