Assay Details
Target Gene Details
Entrez Gene ID: | 55830 |
Gene Name: | glycosyltransferase 8 domain containing 1 |
Gene Aliases: |
AD-017, MSTP139 |
Location: |
Chr.3:52694484-52706083 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLT8D1 | NM_001010983.2 | 5 | 1114 | NP_001010983.1 |
NM_001278280.1 | 5 | 1110 | NP_001265209.1 | |
NM_001278281.1 | 5 | 858 | NP_001265210.1 | |
NM_018446.3 | 4 | 885 | NP_060916.1 | |
NM_152932.2 | 4 | 443 | NP_690909.1 | |
XM_017006857.1 | 6 | 1273 | XP_016862346.1 | |
XM_017006858.1 | 5 | 1044 | XP_016862347.1 | |
XM_017006859.1 | 4 | 593 | XP_016862348.1 | |
AF157318.1 | 4 | 330 | AAF67484.1 | |
AF175227.1 | 5 | 597 | AAQ13652.1 | |
AF176777.1 | 5 | 596 | AAQ13655.1 | |
AJ271710.1 | 3 | 212 | CAC19666.1 | |
AK027517.1 | 4 | 351 | BAB55170.1 | |
AK075031.1 | 3 | 407 | BAC11363.1 | |
AY358579.1 | 4 | 364 | AAQ88942.1 | |
BC119670.1 | 4 | 329 | ||
BC119671.1 | 4 | 329 | ||
BF725274.1 | 5 | 531 | ||
BG700867.1 | ||||
CR749821.1 | 5 | 567 | CAH18681.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |