Assay Details
Target Gene Details
Entrez Gene ID: | 147700 |
Gene Name: | kinesin light chain 3 |
Gene Aliases: |
KLC2, KLC2L, KLCt, KNS2B |
Location: |
Chr.19:45340740-45351520 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLC3 | NM_177417.2 | NP_803136.2 | ||
AK092481.1 | BAC03901.1 | |||
BC062998.1 | ||||
BC073841.1 | AAH73841.1 | |||
BC126418.1 | ||||
BC133037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833845 | Chr.19:45308814 - 45457371 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv29n64 | Chr.19:45334011 - 45373709 on Build GRCh38 | Gain |
![]() ![]() |
esv3644502 | Chr.19:45323147 - 45403933 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv1054e212 | Chr.19:45347119 - 45407663 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv3644504 | Chr.19:45342675 - 45373732 on Build GRCh38 | Gain |
![]() ![]() |
nsv458711 | Chr.19:45323108 - 45408744 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv1063547 | Chr.19:45322368 - 45400204 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
dgv3606n100 | Chr.19:45347435 - 45400204 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv833844 | Chr.19:45181051 - 45384148 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1160648 | Chr.19:45347447 - 45368176 on Build GRCh38 | Duplication |
![]() ![]() |
nsv579868 | Chr.19:45334025 - 45359625 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)