Assay Details
Target Gene Details
Entrez Gene ID: | 79135 |
Gene Name: | apolipoprotein O |
Gene Aliases: |
FAM121B, MIC26, Mic23, My025 |
Location: |
Chr.X:23833348-23907940 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOO | NM_024122.4 | 9 | 999 | NP_077027.1 |
NR_026545.2 | ||||
XM_011545584.2 | 9 | 1308 | XP_011543886.1 | |
XM_011545585.2 | XP_011543887.1 | |||
XM_011545586.2 | XP_011543888.1 | |||
XM_011545587.2 | 8 | 835 | XP_011543889.1 | |
XM_017029837.1 | XP_016885326.1 | |||
AA410995.1 | 1 | 136 | ||
AF061264.1 | AAG43139.1 | |||
AY359114.1 | 9 | 934 | AAQ89472.1 | |
BC002333.2 | 9 | 979 | AAH02333.1 | |
BC010102.2 | 9 | 979 | AAH10102.1 | |
BC016814.1 | 9 | 974 | AAH16814.1 | |
EL951748.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv2237e212 | Chr.X:23829599 - 23856492 on Build GRCh38 | Gain | APOO |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |