Assay Details
Target Gene Details
Entrez Gene ID: | 53938 |
Gene Name: | peptidylprolyl isomerase like 3 |
Gene Aliases: |
CYPJ |
Location: |
Chr.2:200870907-200889333 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPIL3 | NM_032472.3 | 7 | 1048 | NP_115861.1 |
NM_130906.2 | 7 | 1036 | NP_570981.1 | |
XM_005246651.3 | 7 | 967 | XP_005246708.1 | |
XM_005246652.4 | 7 | 955 | XP_005246709.1 | |
XM_011511358.2 | 7 | 1141 | XP_011509660.1 | |
XM_017004352.1 | 7 | 1158 | XP_016859841.1 | |
XM_017004353.1 | 7 | 897 | XP_016859842.1 | |
XM_017004354.1 | 7 | 1331 | XP_016859843.1 | |
XM_017004355.1 | 7 | 1355 | XP_016859844.1 | |
XM_017004356.1 | 7 | 1319 | XP_016859845.1 | |
XM_017004357.1 | 8 | 1238 | XP_016859846.1 | |
XM_017004358.1 | 8 | 1061 | XP_016859847.1 | |
XM_017004359.1 | 8 | 1245 | XP_016859848.1 | |
XM_017004360.1 | 6 | 1058 | XP_016859849.1 | |
AF146799.1 | 7 | 905 | AAO64723.1 | |
AF251049.1 | 7 | 1048 | AAK34939.1 | |
AF271652.1 | 7 | 971 | AAG44766.1 | |
AF414127.1 | 6 | 972 | AAP97286.1 | |
AK297804.1 | ||||
AY257468.1 | 6 | 1092 | AAP13527.1 | |
BC007693.2 | 7 | 1477 | AAH07693.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |