Assay Details
Target Gene Details
Entrez Gene ID: | 441177 |
Gene Name: | long intergenic non-protein coding RNA 602 |
Gene Aliases: |
- |
Location: |
Chr.6:165987551-165989615 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC00602 | NR_027284.1 | 1 | 960 | |
AK094116.1 | 1 | 960 | BAC04291.1 |