Assay Details
Target Gene Details
Entrez Gene ID: | 28972 |
Gene Name: | signal peptidase complex subunit 1 |
Gene Aliases: |
HSPC033, SPC1, SPC12, YJR010C-A |
Location: |
Chr.3:52705841-52708182 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPCS1 | NM_014041.3 | NP_054760.3 | ||
AF092138.1 | AAD40380.1 | |||
AK027426.1 | ||||
AX775783.1 | CAE11591.1 | |||
AX775785.1 | CAE11592.1 | |||
AX775787.1 | CAE11593.1 | |||
AX775833.1 | CAE11616.1 | |||
BC000884.1 | AAH00884.1 | |||
BI826431.1 | ||||
BQ423345.1 | ||||
BU070128.1 | ||||
CA309210.1 | ||||
DB459917.1 | ||||
L38852.1 | AAL31361.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |