Assay Details
Target Gene Details
Entrez Gene ID: | 5250 |
Gene Name: | solute carrier family 25 member 3 |
Gene Aliases: |
OK/SW-cl.48, PHC, PTP |
Location: |
Chr.12:98593625-98602000 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A3 | NM_002635.3 | NP_002626.1 | ||
NM_005888.3 | NP_005879.1 | |||
NM_213611.2 | NP_998776.1 | |||
AB064666.1 | BAB93517.1 | |||
AK057575.1 | ||||
AK074759.1 | BAC11187.1 | |||
AK092689.1 | ||||
AK222658.1 | BAD96378.1 | |||
AK316598.1 | ||||
BC000998.2 | AAH00998.1 | |||
BC001328.2 | AAH01328.1 | |||
BC003504.1 | AAH03504.1 | |||
BC004345.1 | AAH04345.1 | |||
BC006455.1 | AAH06455.1 | |||
BC011574.1 | AAH11574.1 | |||
BC011641.2 | AAH11641.1 | |||
BC014019.2 | AAH14019.1 | |||
BC015379.2 | AAH15379.2 | |||
BC051367.1 | ||||
BX647062.1 | CAE46080.1 | |||
KU178255.1 | ||||
KU178256.1 | ||||
X60036.1 | CAA42641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv559918 | Chr.12:98570959 - 98666572 on Build GRCh38 | Loss | APAF1 SLC25A3 SNORA53 IKBIP |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |