Assay Details
Target Gene Details
Entrez Gene ID: | 64769 |
Gene Name: | MYST/Esa1 associated factor 6 |
Gene Aliases: |
C1orf149, CENP-28, EAF6, NY-SAR-91 |
Location: |
Chr.1:37489960-37514819 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEAF6 | NM_001270875.1 | 7 | 1280 | NP_001257804.1 |
NM_001270876.1 | 6 | 953 | NP_001257805.1 | |
NM_022756.5 | 8 | 1310 | NP_073593.2 | |
NR_073090.1 | 9 | 1368 | ||
NR_073091.1 | 8 | 1338 | ||
NR_073092.1 | 9 | 1399 | ||
AK021792.1 | 7 | 1215 | BAB13898.1 | |
AK225598.1 | 7 | 1215 | ||
AK308118.1 | 8 | 1244 | ||
AK309663.1 | 9 | 1360 | ||
AY007166.1 | 7 | 1205 | ||
BC016328.1 | 7 | 1239 | AAH16328.1 | |
BC056406.1 | 7 | 1236 | AAH56406.1 | |
BX538212.1 | 8 | 1246 | CAD98071.1 | |
BX640719.1 | 8 | 1255 | CAE45838.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv519551 | Chr.1:37491775 - 37509678 on Build GRCh38 | Gain | MIR5581 MEAF6 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |