Assay Details
Target Gene Details
Entrez Gene ID: | 4151 |
Gene Name: | myoglobin |
Gene Aliases: |
PVALB, myoglobgin |
Location: |
Chr.22:35606764-35623354 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MB | NM_005368.2 | 2 | 243 | NP_005359.1 |
NM_203377.1 | 3 | 335 | NP_976311.1 | |
NM_203378.1 | 3 | 318 | NP_976312.1 | |
XM_005261605.2 | 3 | 279 | XP_005261662.1 | |
AK300657.1 | 3 | 201 | ||
AK314057.1 | 3 | 304 | ||
BC014547.1 | 2 | 239 | AAH14547.1 | |
BC018001.1 | 2 | 208 | AAH18001.1 | |
BU585249.1 | 2 | 365 | ||
CB140824.1 | 3 | 348 | ||
CR456516.1 | 2 | 211 | CAG30402.1 | |
CR541949.1 | 2 | 163 | CAG46747.1 | |
HG531366.1 | ||||
HG531368.1 | ||||
HG531369.1 | ||||
HG531372.1 | ||||
HG531373.1 | ||||
HG531378.1 | ||||
HG531379.1 | ||||
HG531380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv510806 | Chr.22:35599609 - 35636220 on Build GRCh38 | Deletion | LOC284912 MB |
nsv834184 | Chr.22:35502044 - 35621639 on Build GRCh38 | Loss | MB RASD2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |