Assay Details
Target Gene Details
Entrez Gene ID: | 5320 |
Gene Name: | phospholipase A2 group IIA |
Gene Aliases: |
MOM1, PLA2, PLA2B, PLA2L, PLA2S, PLAS1, sPLA2 |
Location: |
Chr.1:19975431-19980439 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G2A | NM_000300.3 | 3 | 271 | NP_000291.1 |
NM_001161727.1 | 3 | 240 | NP_001155199.1 | |
NM_001161728.1 | 2 | 160 | NP_001155200.1 | |
NM_001161729.1 | 2 | 194 | NP_001155201.1 | |
AK291302.1 | 2 | 121 | ||
AY656695.1 | AAT73043.1 | |||
AY656696.1 | AAT73044.1 | |||
BC005919.1 | 3 | 248 | AAH05919.1 | |
BM708748.1 | 2 | 137 | ||
BX641714.1 | 2 | 160 | ||
CB132375.1 | 3 | 208 | ||
CR456865.1 | CAG33146.1 | |||
M22430.1 | 2 | 111 | AAA36550.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |