Assay Details
Target Gene Details
Entrez Gene ID: | 10344 |
Gene Name: | C-C motif chemokine ligand 26 |
Gene Aliases: |
IMAC, MIP-4a, MIP-4alpha, SCYA26, TSC-1 |
Location: |
Chr.7:75769524-75791583 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL26 | NM_006072.4 | NP_006063.1 | ||
XM_017011671.1 | 1 | 82 | XP_016867160.1 | |
XM_017011672.1 | XP_016867161.1 | |||
AY358893.1 | AAQ89252.1 | |||
BG211121.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3613737 | Chr.7:75769270 - 75786583 on Build GRCh38 | Gain | CCL26 |
dgv6468n100 | Chr.7:75701432 - 75777782 on Build GRCh38 | Gain | CCL26 HIP1 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | POR MIR4651 HSPB1 LOC100133091 STAG3L1 RHBDD2 LOC107986711 GTF2IP1 LOC441259 TRIM73 STYXL1 NCF1C CCL24 TMEM120A HIP1 SNORA14A PMS2P2 PMS2P10 MDH2 POM121C NSUN5P1 SPDYE5 CCL26 GTF2IP7 SPDYE16 LOC541473 POMZP3 LOC105375355 FDPSP2 PMS2P3 DTX2 SRRM3 YWHAG SSC4D GTF2IRD2B ZP3 |
nsv518028 | Chr.7:75539242 - 75825134 on Build GRCh38 | Gain | CCL26 CCL24 HIP1 |
esv3613726 | Chr.7:75699055 - 75796413 on Build GRCh38 | Gain | CCL26 HIP1 |
esv2764051 | Chr.7:75663839 - 75875518 on Build GRCh38 | Gain+Loss | CCL26 CCL24 HIP1 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | POR MIR4651 HSPB1 LOC100133091 RHBDD2 LOC107986711 STYXL1 CCL24 TMEM120A HIP1 SNORA14A MDH2 POM121C SPDYE5 CCL26 GTF2IP7 SPDYE16 POMZP3 LOC105375355 FDPSP2 PMS2P3 DTX2 SRRM3 YWHAG SSC4D ZP3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |