Assay Details
Target Gene Details
Entrez Gene ID: | 64794 |
Gene Name: | DEAD-box helicase 31 |
Gene Aliases: |
PPP1R25 |
Location: |
Chr.9:132592997-132670403 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX31 | NM_001322340.1 | 1 | 134 | NP_001309269.1 |
NM_001322341.1 | 1 | 134 | NP_001309270.1 | |
NM_001322344.1 | 1 | 134 | NP_001309273.1 | |
NM_022779.8 | 1 | 134 | NP_073616.6 | |
NM_138620.1 | 1 | 134 | NP_619526.1 | |
NR_136309.1 | 1 | 134 | ||
XM_006717236.3 | 1 | 136 | XP_006717299.1 | |
XM_011518921.2 | 1 | 136 | XP_011517223.1 | |
XM_011518922.2 | 1 | 136 | XP_011517224.1 | |
AF335567.1 | 1 | 135 | AAQ14888.1 | |
AF335568.1 | 1 | 135 | AAQ14889.1 | |
AF335569.1 | 1 | 135 | AAQ14890.1 | |
AF427339.1 | 1 | 135 | AAL26549.1 | |
AK027484.1 | 1 | 135 | BAB55146.1 |
Target Gene Details
Entrez Gene ID: | 9329 |
Gene Name: | general transcription factor IIIC subunit 4 |
Gene Aliases: |
KAT12, TF3C-delta, TFIII90, TFIIIC290, TFIIIC90, TFIIICDELTA |
Location: |
Chr.9:132670035-132694955 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GTF3C4 | NM_012204.3 | 1 | 234 | NP_036336.2 |
NR_133925.1 | 1 | 234 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |