Assay Details
Target Gene Details
Entrez Gene ID: | 644815 |
Gene Name: | family with sequence similarity 83 member G |
Gene Aliases: |
PAWS1 |
Location: |
Chr.17:18968789-19006144 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM83G | NM_001039999.2 | 5 | 1904 | NP_001035088.2 |
XM_017024953.1 | 5 | 1988 | XP_016880442.1 | |
AK123558.1 | 3 | 2174 | BAC85645.1 | |
BX365956.2 | ||||
BX402942.2 | 1 | 618 | ||
DY655105.1 |
Target Gene Details
Entrez Gene ID: | 125206 |
Gene Name: | solute carrier family 5 member 10 |
Gene Aliases: |
SGLT-5, SGLT5 |
Location: |
Chr.17:18950676-19020691 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC5A10 | NM_001042450.2 | NP_001035915.1 | ||
NM_001270648.1 | NP_001257577.1 | |||
NM_001270649.1 | NP_001257578.1 | |||
NM_001282417.1 | NP_001269346.1 | |||
NM_152351.4 | NP_689564.3 | |||
XM_011523652.1 | XP_011521954.1 | |||
XM_011523653.2 | XP_011521955.1 | |||
XM_017024191.1 | XP_016879680.1 | |||
AK057946.1 | BAB71619.1 | |||
AK298345.1 | ||||
BC034380.1 | ||||
BC039868.1 | AAH39868.1 | |||
BC062617.1 | AAH62617.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv574552 | Chr.17:18926974 - 18987449 on Build GRCh38 | Gain | SLC5A10 FAM83G PRPSAP2 |
esv33572 | Chr.17:18919047 - 19291177 on Build GRCh38 | Gain | SNORD3D SNORD3C EPN2-IT1 FAM83G LOC102724624 SNORD3A GRAP SNORD3B-2 SLC5A10 EPN2 SNORD3B-1 LOC388436 PRPSAP2 GRAPL LOC79999 |
dgv3116n100 | Chr.17:18957985 - 19190235 on Build GRCh38 | Gain | LOC102724624 SNORD3A GRAP SNORD3B-2 SNORD3D SLC5A10 SNORD3B-1 SNORD3C FAM83G GRAPL LOC79999 |
esv2758678 | Chr.17:18927303 - 19288734 on Build GRCh38 | Gain+Loss | SNORD3D SNORD3C EPN2-IT1 FAM83G LOC102724624 SNORD3A GRAP SNORD3B-2 SLC5A10 EPN2 SNORD3B-1 LOC388436 PRPSAP2 GRAPL LOC79999 |
nsv1057803 | Chr.17:18561260 - 19019420 on Build GRCh38 | Loss | FBXW10 LOC101929141 TRIM16L SLC5A10 CCDC144B TVP23B TBC1D28 ZNF286B FAM83G PRPSAP2 FOXO3B |
nsv953842 | Chr.17:18947088 - 19049187 on Build GRCh38 | Deletion | GRAP SLC5A10 FAM83G |
nsv523721 | Chr.17:18659407 - 19014011 on Build GRCh38 | Gain | FBXW10 LOC101929141 TRIM16L SLC5A10 TVP23B ZNF286B FAM83G PRPSAP2 FOXO3B |
nsv833391 | Chr.17:18906887 - 19238887 on Build GRCh38 | Loss | SNORD3D SNORD3C FAM83G LOC102724624 SNORD3A GRAP SNORD3B-2 SLC5A10 EPN2 SNORD3B-1 LOC388436 PRPSAP2 GRAPL LOC79999 |
dgv565e214 | Chr.17:18920166 - 19019319 on Build GRCh38 | Gain | SLC5A10 FAM83G PRPSAP2 |
nsv428336 | Chr.17:18927239 - 19288734 on Build GRCh38 | Gain+Loss | SNORD3D SNORD3C EPN2-IT1 FAM83G LOC102724624 SNORD3A GRAP SNORD3B-2 SLC5A10 EPN2 SNORD3B-1 LOC388436 PRPSAP2 GRAPL LOC79999 |
nsv9504 | Chr.17:18389579 - 19238707 on Build GRCh38 | Gain+Loss | LGALS9C SNORD3D CCDC144B TVP23B KRT16P1 KRT17P2 FBXW10 SNORD3A LOC101929141 SNORD3B-2 KRT17P5 PRPSAP2 GRAPL TRIM16L SNORD3C ZNF286B FAM83G LOC102724624 EVPLL FLJ35934 GRAP SLC5A10 EPN2 SNORD3B-1 TBC1D28 LOC388436 USP32P2 FOXO3B LOC79999 FAM106A |
dgv5491n54 | Chr.17:18921239 - 19014011 on Build GRCh38 | Gain | SLC5A10 FAM83G PRPSAP2 |
nsv470581 | Chr.17:18947699 - 19014011 on Build GRCh38 | Gain | SLC5A10 FAM83G |
esv3572465 | Chr.17:18971291 - 19019193 on Build GRCh38 | Gain | SLC5A10 FAM83G |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |