Assay Details
Target Gene Details
Entrez Gene ID: | 9727 |
Gene Name: | RAB11 family interacting protein 3 |
Gene Aliases: |
CART1, Rab11-FIP3 |
Location: |
Chr.16:425621-522485 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB11FIP3 | NM_014700.3 | NP_055515.1 | ||
XM_005255713.3 | XP_005255770.1 | |||
XM_005255717.3 | XP_005255774.1 | |||
XM_005255718.3 | XP_005255775.1 | |||
XM_011522765.2 | XP_011521067.1 | |||
XM_017023907.1 | XP_016879396.1 | |||
AB014565.1 | ||||
AF395731.1 | AAL12940.1 | |||
BC051360.1 | AAH51360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | MIR3677 METRN MSLN LUC7L TSC2 SYNGR3 MRPL28 C16orf91 PDIA2 ABCA3 IGFALS PKD1 TELO2 NDUFB10 POLR3K NARFL TPSD1 LOC652276 PRR35 WDR24 RHBDF1 TRAF7 MIR6511B1 MIR6859-4 LOC105371184 LOC729652 PRSS27 AXIN1 RAB26 NTN3 LOC105371038 LOC106660606 HAGHL NUBP2 MIR5587 MIR3178 SNHG9 MSRB1 PRR25 TMEM204 DNASE1L2 BRICD5 SOX8 CCDC154 CCDC78 HN1L LMF1-AS1 RPS2 HBQ1 MIR662 IFT140 CLCN7 LOC107984876 RNF151 LOC105371049 SNORA10 STUB1 ZNF598 LINC00235 LOC105371045 JMJD8 TPSB2 RAB40C NME3 SSTR5-AS1 LMF1 CHTF18 LOC107987233 LOC105371046 ECI1 LOC100134368 CEMP1 MIR6768 PIGQ MAPK8IP3 HBA1 RAB11FIP3 HBM GNPTG RHOT2 LOC105371047 CACNA1H SNHG19 GNG13 RGS11 DDX11L10 FLJ42627 FAHD1 MIR6767 MIR940 LOC101929280 C16orf59 SLC9A3R2 METTL26 PTX4 WDR90 LINC00254 EME2 ATP6V0C FBXL16 C1QTNF8 MIR3176 MRPS34 ABCA17P RPL3L MIR1225 SPSB3 TPSG1 MLST8 AMDHD2 HBA2 ERVK13-1 SNORA64 KCTD5 SNORD60 UBE2I PGP CAPN15 E4F1 NME4 HBZ CASKIN1 DECR2 HS3ST6 TBC1D24 NOXO1 NPW MPG FAM173A MIR4717 UNKL SSTR5 WFIKKN1 CCNF GFER RPUSD1 TSR3 SNRNP25 MIR3177 SNORA78 TBL3 MCRIP2 FAM234A CRAMP1 NPRL3 HAGH WASIR2 PRSS29P MIR4516 TMEM8A NHLRC4 MIR3180-5 LOC100287175 MEIOB RNPS1 ARHGDIG NTHL1 PDPK1 BAIAP3 TPSAB1 RPL23AP5 RHBDL1 |
esv3637576 | Chr.16:439034 - 480097 on Build GRCh38 | Gain | RAB11FIP3 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | FAM234A NME4 RAB11FIP3 TMEM8A DECR2 ARHGDIG MRPL28 LOC100134368 RGS11 PDIA2 RPL23AP5 AXIN1 |
nsv526903 | Chr.16:451362 - 810673 on Build GRCh38 | Loss | METRN MSLN MIR5587 METTL26 PRR25 WFIKKN1 WDR90 CCDC78 RPUSD1 FBXL16 MCRIP2 MIR3176 MIR662 NARFL STUB1 PRR35 WDR24 LINC00235 JMJD8 RAB40C CHTF18 LOC107987233 NHLRC4 LOC105371184 PIGQ CAPN15 LOC100287175 RAB11FIP3 LOC105371038 RHOT2 HAGHL GNG13 FAM173A LOC101929280 RHBDL1 |
nsv1700 | Chr.16:457843 - 476640 on Build GRCh38 | Insertion | RAB11FIP3 |
nsv570954 | Chr.16:342461 - 530124 on Build GRCh38 | Loss | NME4 RAB11FIP3 TMEM8A DECR2 MRPL28 LOC100134368 CAPN15 RPL23AP5 LINC00235 AXIN1 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | FAM234A NME4 RAB11FIP3 TMEM8A DECR2 ARHGDIG MRPL28 LOC100134368 RGS11 PDIA2 RPL23AP5 AXIN1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |