Assay Details
Target Gene Details
Entrez Gene ID: | 22934 |
Gene Name: | ribose 5-phosphate isomerase A |
Gene Aliases: |
RPI, RPIAD |
Location: |
Chr.2:88691658-88750935 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RPIA | NM_144563.2 | NP_653164.2 | ||
AY050633.1 | AAK95569.1 | |||
BC015529.1 | ||||
L35035.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470476 | Chr.2:88384248 - 88829483 on Build GRCh38 | Gain | LOC101928371 FOXI3 RPIA TEX37 ANKRD36BP2 EIF2AK3 LOC101928403 MIR4436A |
dgv61n16 | Chr.2:87343737 - 90254660 on Build GRCh38 | Deletion | LOC101927050 LOC101928371 RPIA MIR4436A PLGLB2 IGKV2-26 LINC00152 THNSL2 IGKV2-29 ANKRD36BP2 LOC101928403 LOC730268 KRCC1 IGKV2-24 MIR4435-1 SMYD1 IGKV1D-13 IGKV1-5 IGKV3-20 FOXI3 IGK IGKV3D-15 MIR4780 TEX37 LOC107985911 EIF2AK3 RGPD2 IGKC FABP1 IGKV1D-8 |
dgv6947n54 | Chr.2:87815738 - 88850700 on Build GRCh38 | Gain | LOC101928371 KRCC1 SMYD1 RPIA MIR4436A THNSL2 FOXI3 MIR4780 TEX37 ANKRD36BP2 EIF2AK3 LOC101928403 RGPD2 FABP1 |
nsv1072005 | Chr.2:87360367 - 90256528 on Build GRCh38 | Deletion | LOC101927050 LOC101928371 RPIA MIR4436A PLGLB2 IGKV2-26 LINC00152 THNSL2 IGKV2-29 ANKRD36BP2 LOC101928403 LOC730268 KRCC1 IGKV2-24 MIR4435-1 SMYD1 IGKV1D-13 IGKV1-5 IGKV3-20 FOXI3 IGK IGKV3D-15 MIR4780 TEX37 LOC107985911 EIF2AK3 RGPD2 IGKC FABP1 IGKV1D-8 |
nsv834290 | Chr.2:88686704 - 88894261 on Build GRCh38 | Loss | IGK RPIA ANKRD36BP2 MIR4436A IGKC |
nsv1130752 | Chr.2:87360364 - 90256525 on Build GRCh38 | Deletion | LOC101927050 LOC101928371 RPIA MIR4436A PLGLB2 IGKV2-26 LINC00152 THNSL2 IGKV2-29 ANKRD36BP2 LOC101928403 LOC730268 KRCC1 IGKV2-24 MIR4435-1 SMYD1 IGKV1D-13 IGKV1-5 IGKV3-20 FOXI3 IGK IGKV3D-15 MIR4780 TEX37 LOC107985911 EIF2AK3 RGPD2 IGKC FABP1 IGKV1D-8 |
nsv582384 | Chr.2:87987149 - 88773214 on Build GRCh38 | Loss | LOC101928371 KRCC1 SMYD1 RPIA THNSL2 FOXI3 MIR4780 TEX37 ANKRD36BP2 EIF2AK3 LOC101928403 RGPD2 FABP1 |
nsv582386 | Chr.2:88035188 - 89208087 on Build GRCh38 | Loss | LOC101928371 KRCC1 IGKV2-24 SMYD1 IGKV1-5 RPIA IGKV3-20 MIR4436A IGKV2-26 THNSL2 FOXI3 IGK MIR4780 TEX37 ANKRD36BP2 EIF2AK3 LOC101928403 IGKC FABP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |