Assay Details
Target Gene Details
Entrez Gene ID: | 4595 |
Gene Name: | mutY DNA glycosylase |
Gene Aliases: |
MYH |
Location: |
Chr.1:45329242-45340925 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MUTYH | NM_001048171.1 | NP_001041636.1 | ||
NM_001128425.1 | NP_001121897.1 | |||
NM_001293190.1 | NP_001280119.1 | |||
NM_001293192.1 | NP_001280121.1 | |||
NM_012222.2 | NP_036354.1 | |||
XM_011541503.2 | XP_011539805.1 | |||
XM_011541505.2 | XP_011539807.1 | |||
XM_017001331.1 | XP_016856820.1 | |||
XM_017001332.1 | XP_016856821.1 | |||
XM_017001335.1 | XP_016856824.1 | |||
XM_017001336.1 | XP_016856825.1 | |||
XM_017001338.1 | XP_016856827.1 | |||
XM_017001339.1 | XP_016856828.1 | |||
AB032920.1 | BAA89336.1 | |||
AB032921.1 | BAA89337.1 | |||
AB032922.1 | BAA89338.1 | |||
AB032923.1 | BAA89339.1 | |||
AK293830.1 | ||||
BC003178.1 | AAH03178.1 | |||
BM926983.1 | ||||
CN413115.1 | ||||
KU178203.1 | ||||
KU178204.1 |
Target Gene Details
Entrez Gene ID: | 114034 |
Gene Name: | target of EGR1, member 1 (nuclear) |
Gene Aliases: |
hCaf1z |
Location: |
Chr.1:45339457-45343978 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOE1 | NM_025077.3 | 1 | 589 | NP_079353.3 |
XM_005270412.3 | 1 | 554 | XP_005270469.1 | |
XM_005270413.4 | 1 | 611 | XP_005270470.1 | |
XM_011540569.2 | 1 | 802 | XP_011538871.1 | |
AK024011.1 | 1 | 110 | BAB14774.1 | |
AK093320.1 | ||||
AK293704.1 | 1 | 203 | ||
AK298162.1 | 1 | 47 | ||
AY169960.1 | 1 | 589 | AAN75441.1 | |
BC009364.2 | AAH09364.1 | |||
CR457320.1 | CAG33601.1 | |||
DA887065.1 | 1 | 515 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2750837 | Chr.1:45330443 - 45442591 on Build GRCh38 | Loss |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)