Assay Details
Target Gene Details
Entrez Gene ID: | 100529251 |
Gene Name: | CKLF-CMTM1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.16:66552563-66579135 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CKLF-CMTM1 | NM_001202509.1 | NP_001189438.1 | ||
NM_001204098.1 | NP_001191027.1 | |||
NM_001204099.1 | NP_001191028.1 | |||
BM929895.1 | ||||
FY211334.1 | ||||
FY212695.1 |
Target Gene Details
Entrez Gene ID: | 113540 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 1 |
Gene Aliases: |
CKLFH, CKLFH1, CKLFSF1 |
Location: |
Chr.16:66566391-66579135 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CMTM1 | NM_052999.3 | 2 | 573 | NP_443725.3 |
NM_181268.2 | 2 | 573 | NP_851785.2 | |
NM_181269.2 | 2 | 278 | NP_851786.1 | |
NM_181270.2 | 2 | 278 | NP_851787.1 | |
NM_181271.2 | NP_851788.1 | |||
NM_181272.2 | NP_851789.1 | |||
NM_181283.2 | NP_851800.1 | |||
NM_181296.2 | NP_851813.1 | |||
AF278576.1 | 2 | 193 | AAK73015.1 | |
AF278577.1 | 2 | 193 | AAK73016.1 | |
AY174118.1 | 2 | 282 | AAN73227.1 | |
AY174119.1 | 2 | 282 | AAN73228.1 | |
AY174120.1 | AAN73229.1 | |||
AY174121.1 | AAN73230.1 | |||
AY174122.1 | AAN73231.1 | |||
AY174123.1 | AAN73232.1 | |||
AY174124.1 | AAN73233.1 | |||
AY174125.1 | 2 | 282 | AAN73234.1 | |
AY174126.1 | 2 | 282 | AAN73235.1 | |
AY174127.1 | AAN73236.1 | |||
AY174128.1 | AAN73237.1 | |||
AY174129.1 | AAN73238.1 | |||
AY174130.1 | AAN73239.1 | |||
AY174131.1 | AAN73240.1 | |||
AY174132.1 | AAN73241.1 | |||
AY174133.1 | AAN73242.1 | |||
AY174134.1 | 2 | 273 | AAN73243.1 | |
AY174135.1 | 2 | 273 | AAN73244.1 | |
AY174136.1 | 2 | 273 | AAN73245.1 | |
AY174137.1 | 2 | 273 | AAN73246.1 | |
AY174138.1 | 2 | 273 | AAN73247.1 | |
BC057852.1 | 2 | 573 | AAH57852.2 | |
DQ309766.1 | 2 | 273 | ABC33727.2 | |
DQ355985.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv123n111 | Chr.16:66568129 - 66581930 on Build GRCh38 | Duplication | CKLF-CMTM1 CMTM2 CMTM1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |