Assay Details
Target Gene Details
Entrez Gene ID: | 25917 |
Gene Name: | THUMP domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.3:9362842-9386791 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
THUMPD3 | NM_001114092.1 | 10 | 1695 | NP_001107564.1 |
NM_015453.2 | 10 | 1900 | NP_056268.2 | |
XM_005265021.4 | 10 | 2257 | XP_005265078.1 | |
XM_005265022.4 | 10 | 2460 | XP_005265079.1 | |
XM_005265023.3 | 10 | 1843 | XP_005265080.1 | |
XM_005265024.4 | 10 | 1843 | XP_005265081.1 | |
XM_006713086.3 | 10 | 2499 | XP_006713149.1 | |
XM_011533570.2 | 10 | 1963 | XP_011531872.1 | |
XM_011533571.2 | 8 | 1714 | XP_011531873.1 | |
XM_017006113.1 | 9 | 2141 | XP_016861602.1 | |
XM_017006114.1 | 9 | 2383 | XP_016861603.1 | |
XM_017006115.1 | 9 | 1784 | XP_016861604.1 | |
XM_017006116.1 | 9 | 1770 | XP_016861605.1 | |
XM_017006117.1 | 8 | 2071 | XP_016861606.1 | |
XM_017006118.1 | 8 | 2313 | XP_016861607.1 | |
XM_017006119.1 | 8 | 1700 | XP_016861608.1 | |
AK001680.1 | 10 | 1659 | BAA91832.1 | |
AK023263.1 | 10 | 1881 | BAB14495.1 | |
AL117483.1 | 7 | 1118 | CAB55956.1 | |
BC001622.1 | 10 | 1674 | AAH01622.1 | |
BC010421.1 | 10 | 2231 | AAH10421.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |