Assay Details
Target Gene Details
Entrez Gene ID: | 53635 |
Gene Name: | prostate tumor overexpressed 1 |
Gene Aliases: |
ACID2, PTOV-1 |
Location: |
Chr.19:49850726-49860744 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTOV1 | NM_001305105.1 | NP_001292034.1 | ||
NM_001305108.1 | NP_001292037.1 | |||
NM_017432.4 | NP_059128.2 | |||
NR_130963.1 | ||||
XM_005258998.4 | XP_005259055.1 | |||
XM_006723244.3 | XP_006723307.1 | |||
XM_011527033.1 | XP_011525335.1 | |||
XM_011527034.2 | XP_011525336.1 | |||
XM_011527035.1 | XP_011525337.1 | |||
XM_017026879.1 | XP_016882368.1 | |||
XM_017026880.1 | XP_016882369.1 | |||
AJ617480.1 | CAE84582.1 | |||
AK057817.1 | ||||
AK294366.1 | 5 | 503 | ||
AY358168.1 | AAQ88535.1 | |||
U79287.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | RCN3 PIH1D1 SNAR-C5 MAMSTR SNAR-A6 SIGLEC16 PLA2G4C PRRG2 LOC100287477 VRK3 FUZ GRIN2D SNAR-B2 C19orf68 SNORD32A GFY SNAR-A5 SNAR-D ZNF473 TBC1D17 BAX LOC101059948 DHDH CGB7 FLJ26850 SNAR-C2 CGB2 SNAR-A1 GRWD1 CARD8 LMTK3 SNAR-A13 SNAR-C1 SNAR-G1 CA11 ALDH16A1 NUCB1 BCAT2 SNAR-G2 TMEM143 MED25 CABP5 NOSIP SNAR-A8 ATF5 PRMT1 ADM5 TSKS SNAR-A14 PTOV1-AS2 PPP1R15A SNORD33 CGB5 MIR5088 LOC105447645 LHB LOC105372430 SPACA4 TRPM4 MIR6798 MIR4750 ZNF114 RPL18 KCNA7 MIR6800 CYTH2 RRAS LIN7B HSD17B14 RPS11 NUP62 RUVBL2 FUT1 FCGRT NUCB1-AS1 SCAF1 PPFIA3 SNORD34 PNKP IZUMO1 SULT2B1 EMP3 SNAR-A12 NTF4 SNAR-C4 MIR4324 IRF3 SLC6A16 SNAR-A10 SNORD35A PLEKHA4 LOC101928295 PRR12 FAM83E PTOV1 SNAR-C3 PTH2 DBP CGB8 CCDC155 DKKL1 SEC1P SNAR-A11 FTL TULP2 NTN5 PLA2G4C-AS1 RASIP1 PTOV1-AS1 HRC SNORD35B RPL13A AP2A1 CCDC114 FUT2 C19orf73 IL4I1 AKT1S1 MIR150 SPHK2 SNAR-A9 LIG1 MIR4749 MIR6799 SNRNP70 MIR4751 BCL2L12 SIGLEC11 ELSPBP1 SNAR-A7 CGB1 SNAR-A4 SNAR-B1 CD37 SNAR-A2 CPT1C KCNJ14 CGB3 SLC17A7 SYNGR4 TEAD2 CARD8-AS1 FGF21 KDELR1 BSPH1 SNAR-A3 FLT3LG GYS1 |
nsv526372 | Chr.19:49853907 - 49877601 on Build GRCh38 | Loss | PTOV1-AS2 TBC1D17 MIR4749 PTOV1 AKT1S1 PNKP |
nsv470150 | Chr.19:49618821 - 49856789 on Build GRCh38 | Gain | PTOV1-AS2 AP2A1 PRR12 PTOV1 SCAF1 CPT1C MED25 MIR5088 FUZ PRMT1 MIR4749 MIR6800 MIR6799 PTOV1-AS1 RRAS ADM5 BCL2L12 TSKS IRF3 |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | PTOV1-AS2 TBC1D17 NUP62 AP2A1 PTOV1 IL4I1 AKT1S1 MED25 PNKP FUZ MIR4750 ATF5 MIR4749 MIR6800 MIR6799 MIR4751 PTOV1-AS1 TSKS |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | NUP62 AP2A1 SCAF1 IL4I1 AKT1S1 MED25 PRRG2 PNKP NOSIP FUZ ATF5 PRMT1 MIR4749 MIR6799 MIR4751 ADM5 BCL2L12 TSKS IRF3 PTOV1-AS2 TBC1D17 PRR12 PTOV1 CPT1C MIR5088 MIR4750 MIR6800 PTOV1-AS1 RRAS |
dgv54n68 | Chr.19:49848512 - 50008788 on Build GRCh38 | Loss | PTOV1-AS2 TBC1D17 NUP62 PTOV1 SIGLEC16 IL4I1 AKT1S1 PNKP VRK3 MIR4750 ATF5 MIR4749 MIR4751 PTOV1-AS1 SIGLEC11 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |