Assay Details
Target Gene Details
Entrez Gene ID: | 254528 |
Gene Name: | meiosis specific with OB domains |
Gene Aliases: |
C16orf73, gs129 |
Location: |
Chr.16:1833983-1872178 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEIOB | NM_001163560.2 | 6 | 693 | NP_001157032.1 |
NM_152764.2 | 6 | 616 | NP_689977.2 | |
BC029829.1 | 2 | 218 | AAH29829.1 | |
CD300613.1 | 3 | 371 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833119 | Chr.16:1834308 - 1911484 on Build GRCh38 | Loss | LINC00254 MEIOB FAHD1 HS3ST6 |
nsv571175 | Chr.16:1806439 - 1875278 on Build GRCh38 | Loss | MEIOB FAHD1 HAGH |
nsv1040440 | Chr.16:1811739 - 1866102 on Build GRCh38 | Gain | MEIOB FAHD1 HAGH |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | CACNA1H HBZ MPG DNASE1L2 MIR6767 GNPTG MRPL28 NME3 MCRIP2 MIR3178 RAB40C FAM234A SYNGR3 LOC105371049 MEIOB ZNF598 CHTF18 C1QTNF8 RGS11 AMDHD2 MAPK8IP3 LMF1 LOC107984876 LOC105371047 SNORA78 LOC106660606 ARHGDIG PDIA2 MIR6859-4 TRAF7 TSC2 DDX11L10 TPSD1 MSRB1 CRAMP1 PRSS29P HBQ1 MIR4516 MIR1225 RPUSD1 SNRNP25 NTN3 HBA1 NUBP2 FLJ42627 RPS2 BAIAP3 FBXL16 LINC00254 SSTR5-AS1 FAM173A STUB1 NDUFB10 PGP WDR90 E4F1 RNF151 WASIR2 MRPS34 PDPK1 MIR5587 HAGHL LOC105371046 EME2 TBC1D24 RHOT2 JMJD8 LOC100134368 MIR3180-5 DECR2 SOX8 TELO2 RHBDF1 ABCA17P RPL3L MIR4717 RPL23AP5 RAB26 SNHG19 SNHG9 CAPN15 PRR35 LMF1-AS1 CCDC78 TPSAB1 SLC9A3R2 WFIKKN1 ERVK13-1 BRICD5 PKD1 METTL26 RNPS1 TMEM204 TMEM8A RAB11FIP3 UNKL MIR3177 PRSS27 WDR24 SNORA10 C16orf91 HBM SNORA64 TPSB2 LUC7L MIR3677 GNG13 ABCA3 MIR940 LOC101929280 SNORD60 LOC105371184 LOC105371038 METRN MLST8 LOC100287175 MIR6768 TSR3 NPRL3 POLR3K PIGQ LOC107987233 TBL3 LOC729652 MIR662 LOC105371045 ECI1 C16orf59 MSLN NPW CEMP1 NOXO1 IGFALS PRR25 SSTR5 FAHD1 NHLRC4 HN1L IFT140 HBA2 CCNF LOC652276 SPSB3 NARFL PTX4 CCDC154 ATP6V0C LINC00235 CLCN7 NTHL1 HAGH TPSG1 NME4 RHBDL1 AXIN1 UBE2I MIR6511B1 CASKIN1 GFER MIR3176 HS3ST6 KCTD5 |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss | LOC105371047 NUBP2 HAGH NME3 IGFALS TELO2 CRAMP1 MEIOB FAHD1 TMEM204 HN1L MAPK8IP3 IFT140 MRPS34 SPSB3 LOC105371046 MIR3177 EME2 |
nsv1046474 | Chr.16:1846666 - 1873690 on Build GRCh38 | Loss | MEIOB |
nsv519366 | Chr.16:1851210 - 1961125 on Build GRCh38 | Loss | LINC00254 MSRB1 NDUFB10 MEIOB RPL3L HS3ST6 |
nsv1042986 | Chr.16:1784723 - 1866102 on Build GRCh38 | Loss | MEIOB NUBP2 FAHD1 HAGH IGFALS |
nsv571177 | Chr.16:1817039 - 1897015 on Build GRCh38 | Gain | LINC00254 MEIOB FAHD1 HAGH |
nsv525861 | Chr.16:1853650 - 1858255 on Build GRCh38 | Loss | MEIOB |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |