Assay Details
Target Gene Details
Entrez Gene ID: | 56980 |
Gene Name: | PR domain 10 |
Gene Aliases: |
PFM7 |
Location: |
Chr.11:129899706-130003097 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRDM10 | NM_020228.2 | 22 | 5795 | NP_064613.2 |
NM_199437.1 | 21 | 5783 | NP_955469.1 | |
NM_199438.1 | 18 | 5482 | NP_955470.1 | |
NM_199439.1 | 16 | 5368 | NP_955471.1 | |
XM_011542912.2 | 21 | 6006 | XP_011541214.1 | |
XM_017018023.1 | 17 | 7555 | XP_016873512.1 | |
AB033057.2 | 18 | 5266 | BAA86545.2 | |
AF275817.1 | 18 | 5482 | AAF87243.1 | |
AF503171.1 | 16 | 5299 | AAP30847.1 | |
AL833172.1 | 21 | 5786 | ||
BC112934.1 | 21 | 5775 | AAI12935.1 | |
BX648944.1 | 22 | 5796 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |