Assay Details
Target Gene Details
Entrez Gene ID: | 57212 |
Gene Name: | TP73 antisense RNA 1 |
Gene Aliases: |
KIAA0495, PDAM |
Location: |
Chr.1:3735984-3747373 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TP73-AS1 | NR_033708.1 | |||
NR_033709.1 | ||||
NR_033710.1 | 5 | 2714 | ||
NR_033711.1 | 4 | 3178 | ||
NR_033712.1 | ||||
AB007964.1 | 4 | 3128 | ||
AL133648.1 | 5 | 1967 | CAB63767.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv460061 | Chr.1:3680453 - 3755227 on Build GRCh38 | Loss | CCDC27 TP73-AS1 TP73 |
nsv470687 | Chr.1:3356499 - 3763210 on Build GRCh38 | Loss | PRDM16 CCDC27 TP73-AS1 ARHGEF16 TP73 TPRG1L MIR551A WRAP73 MEGF6 |
nsv1001715 | Chr.1:3627619 - 3769754 on Build GRCh38 | Loss | CCDC27 TP73-AS1 TP73 TPRG1L WRAP73 |
esv3585042 | Chr.1:3585575 - 4260083 on Build GRCh38 | Gain | LINC01134 LINC01346 CCDC27 TP73 MEGF6 SMIM1 CEP104 TP73-AS1 TPRG1L WRAP73 C1orf174 LRRC47 DFFB |
nsv1012423 | Chr.1:3477686 - 4747449 on Build GRCh38 | Gain | LINC01134 LINC01346 CCDC27 ARHGEF16 TP73 AJAP1 MEGF6 SMIM1 CEP104 TP73-AS1 TPRG1L MIR551A WRAP73 C1orf174 LRRC47 LOC284661 DFFB |
nsv951959 | Chr.1:3051037 - 3812036 on Build GRCh38 | Deletion | MIR4251 CCDC27 ARHGEF16 LINC00982 TP73 LOC105378604 MEGF6 SMIM1 PRDM16 TP73-AS1 TPRG1L MIR551A WRAP73 LRRC47 |
nsv830314 | Chr.1:3660776 - 3804275 on Build GRCh38 | Gain+Loss | SMIM1 CCDC27 TP73-AS1 TP73 LRRC47 |
dgv13n100 | Chr.1:3730448 - 3791316 on Build GRCh38 | Gain | SMIM1 CCDC27 TP73-AS1 TP73 LRRC47 |