Assay Details
Target Gene Details
Entrez Gene ID: | 9637 |
Gene Name: | fasciculation and elongation protein zeta 2 |
Gene Aliases: |
HUM3CL |
Location: |
Chr.2:36552239-36598190 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FEZ2 | NM_001042548.1 | 9 | 1336 | NP_001036013.1 |
NM_005102.2 | 8 | 1255 | NP_005093.2 | |
XM_006712153.2 | 9 | 1158 | XP_006712216.1 | |
AB076184.1 | 8 | 1247 | BAD06207.1 | |
AB209719.1 | 7 | 978 | BAD92956.1 | |
AF113124.1 | 8 | 1154 | AAF14865.1 | |
AK025274.1 | 7 | 935 | ||
AK057492.1 | 1 | 1226 | ||
AL117593.1 | 2 | 3737 | ||
BC089390.1 | 9 | 1282 | AAH89390.1 | |
BC110386.1 | 8 | 1210 | ||
BX537785.1 | 9 | 1181 | CAD97842.1 | |
U60061.1 | 5 | 608 | AAC51283.1 | |
U69140.1 | AAB40661.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |