Assay Details
Target Gene Details
Entrez Gene ID: | 375298 |
Gene Name: | ceramide kinase like |
Gene Aliases: |
RP26 |
Location: |
Chr.2:181536672-181657107 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CERKL | NM_001030311.2 | 14 | 2531 | NP_001025482.1 |
NM_001030312.2 | 10 | 2114 | NP_001025483.1 | |
NM_001030313.2 | 11 | 2246 | NP_001025484.1 | |
NM_001160277.1 | 13 | 2399 | NP_001153749.1 | |
NM_201548.4 | 13 | 2453 | NP_963842.1 | |
NR_027689.1 | 11 | 2257 | ||
NR_027690.1 | 12 | 2389 | ||
AJ640141.1 | 13 | 2370 | CAG26695.1 | |
AK129976.1 | 10 | 1696 | BAC85266.1 | |
AK293844.1 | ||||
BC137498.1 | 13 | 2379 | ||
BC137499.1 | 13 | 2379 |
Target Gene Details
Entrez Gene ID: | 3676 |
Gene Name: | integrin subunit alpha 4 |
Gene Aliases: |
CD49D, IA4 |
Location: |
Chr.2:181456892-181538928 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ITGA4 | NM_000885.5 | 28 | 5766 | NP_000876.3 |
AB208885.1 | 25 | 4544 | BAD92122.1 | |
BC080190.1 | 11 | 3111 | AAH80190.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |