Assay Details
Target Gene Details
Entrez Gene ID: | 7525 |
Gene Name: | YES proto-oncogene 1, Src family tyrosine kinase |
Gene Aliases: |
HsT441, P61-YES, Yes, c-yes |
Location: |
Chr.18:721592-812326 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YES1 | NM_005433.3 | NP_005424.1 | ||
XM_005258139.3 | XP_005258196.1 | |||
XM_017025960.1 | XP_016881449.1 | |||
AK314063.1 | ||||
BC048960.1 | AAH48960.1 | |||
M15990.1 | AAA35735.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv576236 | Chr.18:319176 - 794272 on Build GRCh38 | Gain | COLEC12 ENOSF1 YES1 TYMSOS CLUL1 LOC105376854 TYMS CETN1 |
dgv5810n54 | Chr.18:351004 - 2238246 on Build GRCh38 | Gain | ADCYAP1 COLEC12 ENOSF1 YES1 LINC00470 TYMSOS CLUL1 LOC105376854 TYMS CETN1 LOC388456 |
nsv576242 | Chr.18:567127 - 841320 on Build GRCh38 | Gain | ENOSF1 YES1 TYMSOS CLUL1 TYMS CETN1 |
nsv528169 | Chr.18:551688 - 2240220 on Build GRCh38 | Loss | ADCYAP1 ENOSF1 YES1 LINC00470 TYMSOS CLUL1 TYMS CETN1 LOC388456 |
nsv1060188 | Chr.18:575087 - 863351 on Build GRCh38 | Gain | ENOSF1 YES1 TYMSOS CLUL1 TYMS CETN1 |
esv2761984 | Chr.18:724222 - 855263 on Build GRCh38 | Loss | YES1 |
nsv482935 | Chr.18:10001 - 2900002 on Build GRCh38 | Loss | COLEC12 ENOSF1 YES1 NDC80 USP14 LOC101926997 CETN1 LOC388456 EMILIN2 ADCYAP1 SMCHD1 ROCK1P1 LINC00470 METTL4 CBX3P2 TYMSOS CLUL1 MIR8078 LOC105376854 THOC1 TYMS |
nsv576228 | Chr.18:69836 - 1506323 on Build GRCh38 | Loss | COLEC12 ENOSF1 YES1 USP14 CETN1 LOC388456 ADCYAP1 ROCK1P1 LINC00470 TYMSOS CLUL1 MIR8078 LOC105376854 THOC1 TYMS |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |