Assay Details
Target Gene Details
Entrez Gene ID: | 3980 |
Gene Name: | DNA ligase 3 |
Gene Aliases: |
LIG2 |
Location: |
Chr.17:34980498-35009169 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIG3 | NM_002311.4 | 2 | 200 | NP_002302.2 |
NM_013975.3 | 2 | 200 | NP_039269.2 | |
XM_005257970.3 | 2 | 168 | XP_005258027.1 | |
XM_006721896.3 | 2 | 169 | XP_006721959.1 | |
XM_017024624.1 | 2 | 364 | XP_016880113.1 | |
AK300363.1 | 2 | 168 | ||
AK308407.1 | 2 | 145 | ||
BC009026.1 | 2 | 121 | AAH09026.1 | |
BC068005.1 | 2 | 175 | AAH68005.1 | |
BM759573.1 | 2 | 179 | ||
DB100562.1 | 2 | 200 | ||
U40671.1 | 2 | 161 | AAA85022.1 | |
X84740.1 | 2 | 143 | CAA59230.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |