Assay Details
Target Gene Details
Entrez Gene ID: | 9585 |
Gene Name: | kinesin family member 20B |
Gene Aliases: |
CT90, KRMP1, MPHOSPH1, MPP-1, MPP1 |
Location: |
Chr.10:89701590-89774943 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KIF20B | NM_001284259.1 | 11 | 1292 | NP_001271188.1 |
NM_016195.3 | 11 | 1292 | NP_057279.2 | |
AB033337.1 | 11 | 1245 | BAB69456.1 | |
AK074336.1 | 11 | 1243 | ||
AK290439.1 | 11 | 1272 | ||
AL117496.1 | 11 | 1247 | CAB55962.1 | |
AY282406.1 | 11 | 1269 | AAP40330.1 | |
BC046134.1 | 11 | 1245 | AAH46134.1 | |
BC058913.1 | 11 | 1248 | AAH58913.1 | |
BC093089.1 | 11 | 1247 | AAH93089.1 | |
BC108688.1 | 11 | 1254 | AAI08689.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1054572 | Chr.10:89712199 - 89777313 on Build GRCh38 | Loss | KIF20B |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |